Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PRMT9 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA548942
Description
PRMT9 Polyclonal Antibody for Western Blot
Substrate recognition component of a SCF (SKP1-CUL1-F-box protein) E3 ubiquitin-protein ligase complex which mediates the ubiquitination and subsequent proteasomal degradation of target proteins, such as DTL/CDT2, BCL6 and PRDM1/BLIMP1. The SCF(FBXO11) complex mediates ubiquitination and degradation of BCL6, thereby playing a role in the germinal center B-cells terminal differentiation toward memory B-cells and plasma cells. The SCF(FBXO11) complex also mediates ubiquitination and degradation of DTL, an important step for the regulation of TGF-beta signaling, cell migration and the timing of the cell-cycle progression and exit. Binds to and neddylates phosphorylated p53/TP53, inhibiting its transcriptional activity. SCF(FBXO11) does not seem to direct ubiquitination of p53/TP53.
Specifications
PRMT9 | |
Polyclonal | |
Unconjugated | |
PRMT9 | |
AI931714; Prmt10; PRMT9; protein arginine methyltransferase 10 (putative); protein arginine methyltransferase 9; Protein arginine N-methyltransferase 10; Protein arginine N-methyltransferase 9; putative protein arginine N-methyltransferase 10; putative protein arginine N-methyltransferase 9; RGD1306157 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
102182, 90826 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q3U3W5, Q6P2P2 | |
PRMT9 | |
KLH conjugated synthetic peptide between 39-68 amino acids from the N-terminal region of human PRMT10. | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction