Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PRNT Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA583752
Description
Immunogen sequence: LDASIHPFRL PFSSKPFLLI PMSNTTLPHT AWPLSFLHQT VSTLKAVAVT HSLWHLQIPV DCQACNR Highest antigen sequence indentity to the following orthologs: Mouse - 27%, Rat - 28%.
PRNT is a protein coding gene. Gene ontology (GO) annotation include extracellular region/cellular component.
Specifications
| PRNT | |
| Polyclonal | |
| Unconjugated | |
| PRNT | |
| M8; prion protein (testis specific); PRNT | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 149830 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q86SH4 | |
| PRNT | |
| Recombinant protein corresponding to Human PRNT. Recombinant protein control fragment (Product #RP-99458). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction