Learn More
Invitrogen™ pro-Caspase 7 Recombinant Rabbit Monoclonal Antibody (SD20-57)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA532355
Description
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

Specifications
pro-Caspase 7 | |
Recombinant Monoclonal | |
1 mg/mL | |
TBS with 0.05% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4 | |
P55210 | |
CASP7 | |
Synthetic peptide within Human Caspase 7 aa 1-50. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot | |
SD20-57 | |
Unconjugated | |
CASP7 | |
Apoptotic protease Mch-3; CASP7; CASP-7; caspase 7; caspase 7, apoptosis-related cysteine peptidase; caspase 7, apoptosis-related cysteine protease; caspase-7; Caspase-7 subunit p11; Caspase-7 subunit p20; CMH-1; ICE-LAP3; ICE-like apoptotic protease 3; LICE2; MCH3 | |
Rabbit | |
Protein A | |
RUO | |
840 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.