Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ pro-EGF Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA529229
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA529229 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA529229 Supplier Invitrogen™ Supplier No. PA529229
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted reactivity: Rat (80%), Dog (85%), Cat (86%), Pig (85%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Epidermal growth factor has a profound effect on the differentiation of specific cells in vivo and is a potent mitogenic factor for a variety of cultured cells of both ectodermal and mesodermal origin. The EGF precursor is believed to exist as a membrane-bound molecule which is proteolytically cleaved to generate the 53-amino acid peptide hormone that stimulates cells to divide.
TRUSTED_SUSTAINABILITY

Specifications

Antigen pro-EGF
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene EGF
Gene Accession No. P01133, P07522
Gene Alias beta-urogastrone; Egf; Epidermal growth factor; HOMG4; Pro-epidermal growth factor; URG; Urogastrone
Gene Symbols EGF
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the Extracellular domain of human pro-EGF. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1950, 25313
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.