Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pro-Lab Diagnostics™ Microbank™ Bacterial and Fungal Preservation System
SDP

Catalog No. p-3770363 Shop All Pro-Lab Diagnostics Products
Click to view available options
Closure Color:
Assorted
Blue
Green
Light Blue
Red
Yellow
This item is not returnable. View return policy
This item is not returnable. View return policy

Designed to simplify the storage and retrieval of bacterial and fungal cultures. Prolab Diagnostics™ Microbank™ Bacterial and Fungal Preservation System is a ready-to-use system.

  • Incorporates treated beads and a special cryopreservative solution
  • Each vial contains 25 beads, providing repeated identical cultures from the original organism
  • Specially formulated preservative ensures longer survival of fastidious cultures and higher quantitative recoveries
  • 80 Vials supplied in a freezer box with“see through”cover
  • Available in 5 cap/bead colors or mixed (16 vials each of blue, green, light blue, red, and yellow)

Order Info

System supplied as box of 80 vials

Specifications

Product Type Cryogenic Storage Vial
Closure Color Green
For Use With (Application) Designed to simplify the storage and retrieval of bacterial and fungal cultures
Quantity 80 Pk.
Storage Requirements -70°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.