Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pro-Lab Diagnostics™ Prolex™ - Blue E. coli 0157 Latex Test Reagent Kits

Click to view available options
Quantity:
100 Tests
50 Tests
Description
- Developed to remove the possibility of cross-reactions with E. hermanii and other sorbitol negative Escherichia species
- Latex agglutination
- Test results in 2 minutes
- CLIA Complexity: Non-waived
- Sensitivity of 100%
Specifications
Specifications
Content And Storage | 2°C to 8°C |
Clia Complexity | Non-waived |
Detectable Analytes | E. coli serogroup O157 |
Sensitivity | 1 |
Type | Blue E. coli O157 Latex Test Reagent Kit |
Includes | E. coli O157 Latex reagent 6.2mL, negative control Latex reagent 3mL, positive control 3mL, Agglutination cards 50, mixing sticks 150 |
Quantity | 100 Tests |
Test Time | 2 min. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction