Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pro-Lab Diagnostics™ Prolex™ Complete Streptococcal Grouping Latex Kit
SDP

Catalog No. 22286175 Shop All Pro-Lab Diagnostics Products
Click to view available options
Quantity:
60 Tests
This item is not returnable. View return policy

Catalog No. 22-286-175

Supplier: Pro-Lab Diagnostics™ PL.030

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Includes

Specific Latex Suspensions for Groups A-G (3mL each); Polyvalent Positive Control (2mL); Extraction Reagent 1 (3.2mL); Extraction Reagent 2 (3.2mL); Extraction Reagent 3 (2 x 8.0mL); Agglutination Cards (48); Mixing Sticks (350)

Requires

  • Inoculating loops
  • Pasteur pipets
  • Borosilicate glass test tubes (12mm x 75mm)
  • Timer

Provides a rapid method for the serological identification of groups A, B, C, D, F and G of the Lancefield groups of streptococci grown on agar plates

  • Offers a unique method for grouping Lancefield beta-haemolytic Streptococci
  • Utilizes modified acid extraction reagents and blue carboxylated latex particles
  • Results are obtained within one minute at room temperature
  • All kit reagent labels and caps are color coded for ease of identification, and presented in easy-to-use, convenient dropper bottles

All components are also available individually

Specifications

Content And Storage 2°C to 8°C
Sensitivity 99%
Type Complete Streptococcal Grouping Latex Kit
Includes 3mL Specific Latex suspensions for groups A, B, C, D, F and G, 2mL Polyvalent positive control, 3.2mL extraction R1, 3.2mL extraction R2, 2 x 8mL extraction R3, 48 Agglutination cards and 350 mixing sticks
Quantity 60 Tests
Specificity 1
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.