Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pro-Lab Diagnostics™ Prolex™ Streptococcal Grouping Latex Kits
Shop All Pro-Lab Diagnostics Products

Click to view available options
Quantity:
2 mL
3 mL
Type:
Group A Latex Reagent
Group B Latex Reagent
Group C Latex Reagent
Group D Latex Reagent
Group F Latex Reagent
Group G Latex Reagent
Polyvalent Positive Control
Description
- Instant results obtained at room temperature
- Advanced extraction process using modified nitrous acid reagents
- Enhanced results with blue carboxylate-modified microparticles (CM-MP)
- As few as one colony required
- Colour coded reagents and vials for ease of selection
- Available individually
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Type | Group F Latex Reagent |
| Includes | 3mL Blue Latex particles coated with purified Rabbit Antibodies |
| Quantity | 3 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction