Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ pro-Orexin, Mouse, Unlabeled, Clone: 3H9.3,

Catalog No. MABN1526MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABN1526MI 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN1526MI Supplier MilliporeSigma™ Supplier No. MABN1526
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Orexin (UniProt O43612; also known as Hcrt, Hypocretin) is encoded by the HCRT (also known as NRCLP1, OX, PPORX, and PPOX) gene (Gene ID 3060) in human. Orexins are highly conserved excitatory neuropeptides that serve as endogenous ligands for an orphan G-protein-coupled receptor (GPCR) and play a significant role in the regulation of food intake and sleep-wakefulness cycle. Orexin-A and -B are derived from the same precursor peptide (prepro-orexin) by proteolytic processing. They exhibit about 50% sequence identity. Orexin-A consists of 33 amino acid residues with two intra-chain disulfide bonds and orexin-B consists of a linear chain of 28 amino acids. They bind and activate two closely related GPCRs, termed OX1 and OX2 receptors. OX1 appears to be selective for orexin-A, whereas OX2 is a nonselective receptor and binds both orexin-A and -B. Orexins are predominantly localized in neurons located in the lateral hypothalamic area, a region implicated in feeding behavior. However, orexin-containing neurons diffusely innervate the entire brain, including monosynaptic projections, to various regions of the cerebral cortex, limbic system, and brain stem. Deficiency of orexin is reported to be a causative factor in human narcolepsy. Ref.: Martynska, L., et al. (2005). Neuroendocrinol. Lett. 26(4):289-292
TRUSTED_SUSTAINABILITY

Specifications

Antigen pro-Orexin
Applications Western Blot
Classification Monoclonal
Clone 3H9.3
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse IgG2aκ in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene HCRT, NRCLP1, OX, PPORX, PPOX
Gene Accession No. O43612
Host Species Mouse
Immunogen GST-tagged recombinant human orexin fragment composed of orexin-B and the propeptide region sequence.
Purification Method Protein G Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_001515
Test Specificity Clone 3H9.3 epitope resides within the propeptide sequence and will detect prepro- and pro-orexin, but not mature orexin, orexin A, or orexin B.
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.