Learn More
Gilson™ Pro Pipette Service for Multichannel Pipettes
GilsonTM Pro Pipette Service offers quality service, preventative maintenance, and calibration to ensure pipettes lasts for years to come.
Supplier:  Gilson™ SVC101140
 View more versions of this product
Catalog No. SVC101140G
Only null left
Add to Cart
Description
Description
  • Visual and Functional Inspection
  • Internal and External Cleaning
  • Leak Test
  • Gravimetric Check
  • Preventative Maintenance
  • Re-grease as needed
  • Pipette Label
  • Mail-In or Onsite Service Available
  • Calibration and Detailed Certificate
  • ISO 17025 Calibration Available
  • 5 day turn around time
Specifications
Specifications
Multi-channel Pipette
Data collected before and after repair
ISO Pipette Pro PM and Calibration Service
ISO 17025 Accredited Service
5 Days
AF/AR; 4x2
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.