Learn More
Gilson™ Pro Pipette Service for Multichannel Pipettes
Gilson Pro Pipette Service offers quality service, preventative maintenance, and calibration to ensure pipettes lasts for years to come.
Supplier: Gilson™ SVC101152
Description
- Visual and Functional Inspection
- Internal and External Cleaning
- Leak Test
- Gravimetric Check
- Preventative Maintenance
- Re-grease as needed
- Pipette Label
- Mail-In or Onsite Service Available
- Calibration and Detailed Certificate
- ISO 17025 Calibration Available
- 5 day turn around time
Specifications
| Multi-channel Pipette | |
| Data collected before and after repair | |
| 5 Days | |
| As-found/After Repair Calibration |
| ISO 17025 Accredited Service | |
| 5 x 3 Measurements | |
| Pipette Pro PM and Calibration Service |