Description
- Visual and Functional Inspection
- Internal and External Cleaning
- Leak Test
- Gravimetric Check
- Preventative Maintenance
- Re-grease as needed
- Pipette Label
- Mail-In or Onsite Service Available
- Calibration and Detailed Certificate
- ISO 17025 Calibration Available
- 5 day turn around time
Specifications
Specifications
| For Use With (Equipment) | Single-channel Pipette |
| Certifications/Compliance | ISO 17025 Accredited Service |
| Description | Data collected after repair |
| Calibration Points | 4 x 2 Measurements |
| Duration | 5 Days |
| Calibration Type | Gravimetric |
| Product Type | Preventive Maintenance and Calibration |
| Product Line | Pipette Pro |
| Service Offering | After Repair Calibration |