Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Pro-Q™ Diamond Phosphoprotein Gel Staining Kits

Catalog No. p-7229245
Encompass_Preferred
Click to view available options
Quantity:
1 L Stain, 40 μL Standard Kit
200 mL Stain, 40 μL Standard Kit

The Pro-Q Diamond Phosphoprotein Gel Staining Kit includes Pro-Q Diamond Phosphoprotein Gel Stain and PeppermintStick Phosphoprotein Molecular Weight Standards.

The Pro-Q Diamond Phosphoprotein Gel Staining Kit includes Pro-Q Diamond Phosphoprotein Gel Stain and PeppermintStick Phosphoprotein Molecular Weight Standards. Pro-Q Diamond Phosphoprotein Gel Stain is a fluorescent stain (excitation/emission maxima ∼555/580 nm) that can detect as little as 1–16 ng of phosphoprotein per band with a signal that is linear over three orders of magnitude. The PeppermintStick Phosphoprotein Molecular Weight Standards are a mixture of phosphorylated and nonphosphorylated proteins with molecular weights from 14,000 to 116,250 daltons that serve both as molecular weight markers and as positive and negative controls.

The Pro-Q Diamond Phosphoprotein Gel Staining Kit includes Pro-Q Diamond Phosphoprotein Gel Stain and PeppermintStick Phosphoprotein Molecular Weight Standards. Pro-Q Diamond Phosphoprotein Gel Stain is a fluorescent stain (excitation/emission maxima ∼555/580 nm) that can detect as little as 1–16 ng of phosphoprotein per band with a signal that is linear over three orders of magnitude. PeppermintStick Phosphoprotein Molecular Weight Standards are a mixture of phosphorylated and nonphosphorylated proteins with molecular weights from 14,000 to 116,250 daltons that serve both as molecular weight markers and as positive and negative controls.

Features include:
Sensitive—detection level down to 1–16 ng of phosphoprotein per band
Convenient—selectively stain phosphoproteins in acrylamide gels, without the need for blotting or phosphoprotein-specific antibodies and western blot analysis
Built-in controls—molecular weight marker allows for approximation of protein size and serves as positive and negative control with unique mixture of phosphorylated and non-phosphorylated proteins
Linear signal—exhibits excellent linearity over three orders of magnitude, ideal for quantitation

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • Pro-Q™ Diamond Phosphoprotein Gel Stain, 200 mL
•PeppermintStick™ Phosphoprotein Molecular Weight Standard, 40 μL
Description Pro-Q™ Diamond Phosphoprotein Gel Staining Kit, 200 mL
Detection Location In-Gel Detection
Detection Method Fluorescence
Label or Dye Pro-Q Diamond
Product Type Phosphoprotein Gel Staining Kit
Product Line PRO-Q™
Quantity 200 mL Stain, 40 μL Standard Kit
Target Molecule Proteins (Phosphoproteins)
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.