Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ PRO Series NightHawk™ 5555 Structural Boot: Extra-Wide Width

Pull-on firefighter boot provides the high-traction, flame-resistant and slip-resistant structural firefighting security you need in the fireground. Honeywell PRO Series NightHawk™ 5555 Structural Boot: Extra-Wide Width combines durability, protection, and support with world class dexterity.

$633.00 - $633.50

Specifications

Certifications/Compliance NFPA 1971-2013 - Structural FF and NFPA 1992-2012 - Liquid Splash
Color Black
For Use With (Application) Structural firefighting
Heel Style Beveled
Liner Material CROSSTECH™ Footwear Fabric / Cambrelle™ Lining
View More Specs

Products 23
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-300-312
View Documents
Honeywell™
BT5555050XW
5
Pair for $633.00
Only null left
Add to Cart
 
19-300-315
View Documents
Honeywell™
BT5555055XW
5.5
Pair for $633.00
Only null left
Add to Cart
 
19-300-318
View Documents
Honeywell™
BT5555060XW
6
Pair for $633.00
Only null left
Add to Cart
 
19-300-321
View Documents
Honeywell™
BT5555065XW
6.5
Pair for $633.00
Only null left
Add to Cart
 
19-300-324
View Documents
Honeywell™
BT5555070XW
7
Pair for $633.00
Only null left
Add to Cart
 
19-300-327
View Documents
Honeywell™
BT5555075XW
7.5
Pair for $633.00
Only null left
Add to Cart
 
19-300-330
View Documents
Honeywell™
BT5555080XW
8
Pair for $633.00
Only null left
Add to Cart
 
19-300-333
View Documents
Honeywell™
BT5555085XW
8.5
Pair for $633.00
Only null left
Add to Cart
 
19-300-336
View Documents
Honeywell™
BT5555090XW
9
Pair for $633.00
Only null left
Add to Cart
 
19-300-345
View Documents
Honeywell™
BT5555105XW
10.5
Pair for $633.00
Only null left
Add to Cart
 
19-300-342
View Documents
Honeywell™
BT5555100XW
10
Pair for $633.00
Only null left
Add to Cart
 
19-300-339
View Documents
Honeywell™
BT5555095XW
9.5
Pair for $633.00
Only null left
Add to Cart
 
19-300-348
View Documents
Honeywell™
BT5555110XW
11
Pair for $633.00
Only null left
Add to Cart
 
19-300-351
View Documents
Honeywell™
BT5555115XW
11.5
Pair for $633.00
Only null left
Add to Cart
 
19-300-354
View Documents
Honeywell™
BT5555120XW
12
Pair for $633.00
Only null left
Add to Cart
 
19-300-357
View Documents
Honeywell™
BT5555125XW
12.5
Pair for $633.00
Only null left
Add to Cart
 
19-300-360
View Documents
Honeywell™
BT5555130XW
13
Pair for $633.50
Only null left
Add to Cart
 
19-300-363
View Documents
Honeywell™
BT5555135XW
13.5
Pair for $633.50
Only null left
Add to Cart
 
19-300-369
View Documents
Honeywell™
BT5555145XW
14.5
Pair for $633.00
Only null left
Add to Cart
 
19-300-366
View Documents
Honeywell™
BT5555140XW
14
Pair for $633.00
Only null left
Add to Cart
 
19-300-372
View Documents
Honeywell™
BT5555150XW
15
Pair for $633.00
Only null left
Add to Cart
 
19-300-375
View Documents
Honeywell™
BT5555155XW
15.5
Pair for $633.00
Only null left
Add to Cart
 
19-300-378
View Documents
Honeywell™
BT5555160XW
16
Pair for $633.00
Only null left
Add to Cart
 
Description

Description

  • FR Rubber Shin Guard: Ladder protection with cushioned backing for utility and comfort
  • Graded Calf Size: Shaft opening increases with foot size– virtually eliminates need for expensive wide calf model
  • Heavyweight Kevlar™: Premium thread to maximize seam strength and durability
  • Premium Full-Grain Cowhide Leather: Flame-resistant, water-resistant, leather upper; flexible joint transitions for world-class comfort
  • PowerToe Protection: Our updated innovation for outstanding abrasion resistant durability
  • Oblique Steel Toe Cap: A roomy fit without unnecessary bulk
  • Vibram™ Racer Outsole: Beveled heel and notched perimeter lugs to move over uneven terrain with confidence. Omni-directional tread pattern with Self-cleaning lugs for great traction
  • Leather-Reinforced Nomex™ Pull Straps: Easy grip, and kind to pants and legs
  • CROSSTECH™ Footwear Fabric / Cambrelle™ Lining: Full-height protection and comfort from underfoot to the boot-top line
  • Leather Top Binding: Always a smooth, finished interface for pants and legs
  • Heel and Ankle Grip System: Molded heel counter, contoured, lined, integrated cushion support for outstanding fit and comfort
  • Heel Kicker: High visibility doffing aid
  • 3M Scotchlite™ Reflective Trim: Located below the pant line for maximum benefit. Cleanable and heat resistant
  • Rugged, Reliable Protection: Steel triple-ladder shank and steel flex-sole plate. Puncture and penetration resistance
  • Deep Cup Outsole: Premium cement, insensitive to temperature, connects the upper to the outsole. The cup wall depth enhances support and durability of the leather upper while maintaining great flexibility
Specifications

Specifications

NFPA 1971-2013 - Structural FF and NFPA 1992-2012 - Liquid Splash
Structural firefighting
CROSSTECH™ Footwear Fabric / Cambrelle™ Lining
Leather
Triple ladder
Steel
Oblique Steel
Omni-directional
Black
Beveled
Notched perimeter
Steel
Puncture and penetration resistant plate
Leather-Reinforced Nomex™
Oblique Steel Toe Cap
Extra-Wide Structural boot
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.