Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PROB1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563816
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563816 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563816 Supplier Invitrogen™ Supplier No. PA563816
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VKTTYAPGFP AGAQGSGLPA PPADPCGEEG GESKTQEPPA LGPPAPAHYT SVFIKDFLPV VPHPYEPPEP S Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 82%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen PROB1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PROB1
Gene Accession No. E7EW31
Gene Alias C5orf65; PROB1; proline rich basic protein 1; proline-rich basic protein 1; weakly similar to basic proline-rich protein
Gene Symbols PROB1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PROB1. Recombinant protein control fragment (Product #RP-104172).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 389333
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.