Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
proBDNF Antibody - Azide and BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $501.50
Specifications
Antigen | proBDNF |
---|---|
Concentration | LYOPH |
Applications | Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017947
![]() |
Novus Biologicals
NB100987560.1MG |
0.1 mg |
Each for $206.50
|
|
|||||
NB10098756
![]() |
Novus Biologicals
NB10098756 |
0.5 mg |
Each for $501.50
|
|
|||||
Description
proBDNF Polyclonal specifically detects proBDNF in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.Specifications
proBDNF | |
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Cytokine Research, Immunology, Neuroscience | |
Sterilized water with No Preservative | |
abrineurin, brain-derived neurotrophic factor, MGC34632, neurotrophin | |
BDNF | |
Primary | |
proBDNF. |
LYOPH | |
Polyclonal | |
Purified | |
RUO | |
Rat, Human, Mouse | |
P23560 | |
627 | |
IgG | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title