Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
proBDNF Antibody, Novus Biologicals™

Guinea Pig Polyclonal Antibody
Supplier: Novus Biologicals NBP280497
Description
proBDNF Polyclonal specifically detects proBDNF in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
proBDNF | |
Polyclonal | |
Western Blot 1:1000, Immunohistochemistry 1:1000, Immunohistochemistry-Paraffin 1:1000 | |
abrineurin, brain-derived neurotrophic factor, MGC34632, neurotrophin | |
Guinea Pig | |
Unpurified | |
RUO | |
Primary | |
Reconstitute in 0.1 ml of sterile water. Centrifuge to remove any insoluble material. Glycerol may be added (1:1) for additional stability. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Lyophilized from whole serum | |
BDNF | |
A synthetic peptide from mouse pro BDNF conjugated to blue carrier protein was used as the antigen. | |
0.1 mL | |
Cytokine Research, Immunology, Neuroscience | |
627 | |
Human, Mouse, Rat, Monkey |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction