Learn More
Invitrogen™ proBDNF Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA118374
Description
Reconstitute in 20 μL of sterile water. Centrifuge to remove any insoluble material. After reconstitution keep aliquots at -20 °C for a higher stability, and at 4 °C with an appropriate antibacterial agent. Glycerol (1:1) may be added for an additional stability. Avoid repetitive freeze/thaw cycles.
BDNF (Brain-Derived Neurotrophic Factor) is a member of the neurotrophin family. BDNF is synthesized as pre-proBDNF, followed by cleavage to proBDNF. Although further processing generates the mature, 14 kDa protein, pro-BDNF is biologically active and is secreted in synaptic vesicles along with the mature form. BDNF is widely expressed in the central nervous system and acts in an autocrine and paracrine manner on several classes of neurons. Signaling occurs mainly through the tyrosine kinase receptor TrkB, although binding to the lower-affinity receptor, p75-NTR, has also been demonstrated. BDNF promotes neuronal survival and differentiation and has been shown to play a critical role in memory formation and synaptic regulation. BDNF is induced by cortical neurons, and is necessary for survival of striatal neurons in the brain. Expression of BDNF is reduced in both Alzheimers and Huntington disease patients, and may also play a role in the regulation of stress response and in the biology of mood disorders.
Specifications
| proBDNF | |
| Polyclonal | |
| Unconjugated | |
| Bdnf | |
| abrineurin; ANON2; anorexia BDNF; Bdnf; BDNF precursor form; brain derived neurothrophic factor; brain derived neurotrophic factor; brain-derived neurotrophic factor; BULN2; neurotrophin; pro BDNF; ProBDNF | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 12064, 24225, 627 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Glycerol (1:1) may be added for added stability. | |
| Lyophilized |
| Immunohistochemistry (Frozen), Western Blot | |
| 1 mg/mL | |
| PBS with no preservative | |
| P21237, P23363, P23560 | |
| Bdnf | |
| Synthetic peptide corresponding to residues (C-E(69) L L D E D Q K V R P N E E(82) of human BDNF precursor. | |
| 20 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.