Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
proBDNF Antibody - Azide and BSA Free, Novus Biologicals™

Sheep Polyclonal Antibody
$206.50 - $478.00
Specifications
Antigen | proBDNF |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 10-50 ug/ml Immunohistochemistry 10-50 ug/ml Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 10-50 ug/ml |
Applications | Immunohistochemistry (Paraffin), Immunohistochemistry, Western Blot, Immunofluorescence, Immunocytochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017946
![]() |
Novus Biologicals
NB100987540.1MG |
0.1 mg |
Each for $206.50
|
|
|||||
NB10098754
![]() |
Novus Biologicals
NB10098754 |
0.5 mg |
Each for $478.00
|
|
|||||
Description
proBDNF Polyclonal specifically detects proBDNF in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
proBDNF | |
Western Blot 10-50 ug/ml Immunohistochemistry 10-50 ug/ml Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 10-50 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
Mouse, Rat | |
abrineurin, brain-derived neurotrophic factor, MGC34632, neurotrophin | |
BDNF | |
Primary | |
proBDNF. |
LYOPH | |
Immunohistochemistry (Paraffin), Immunohistochemistry, Western Blot, Immunofluorescence, Immunocytochemistry | |
Unconjugated | |
Sheep | |
Cytokine Research, Immunology, Neuroscience | |
P23560 | |
627 | |
IgG | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title