Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G-Biosciences Probe-Based qPCR 2X Master Mix
SCI_ED

Catalog No. S03135
Change view
Click to view available options
No. of Reactions:
100
500
1000
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
S03135 100
S03133 1000
S03134 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S03135 Supplier G-Biosciences Supplier No. 7861982
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Probe-based PCR 2X master mix is used for gene expression analysis, genotyping and mutation detection, pathogen detection, viral detection, absolute quantification. It has enhanced efficiency, specificity, sensitivity and compatible with nearly all real time PCR instruments.

  • G-biosciences probe-based qPCR 2X master mix contains Taq DNA polymerase, low ROX reference dye, dNTPs and optimized buffer with stabilizers.
  • This master mix is ideal for high-throughput real-time PCR screening and validation which requires minimal handling and offer robust and consistent results.

Specifications

Detection Method Primer-probe
Form Liquid
GC-Rich PCR Performance High
PCR Method 1-step RT-qPCR, 2-step RT-qPCR, Multiplex qPCR, qPCR, RT-qPCR
Reaction Speed Fast or Standard
Product Type qPCR Master Mix
For Use With (Application) Gene Expression, Genotyping, Microbial Detection, Pathogen Detection, Virus Detection
No. of Reactions 100
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.