Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PROCA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569898
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569898 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569898 Supplier Invitrogen™ Supplier No. PA569898
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Human: 100%; Rabbit: 92%.

PROCA1 gene ontology annotations related to this gene include arachidonic acid secretion; phospholipid metabolic process.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PROCA1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PROCA1
Gene Accession No. Q8NCQ7
Gene Alias 4933404M19Rik; PROCA1; proline-rich cyclin A1-interacting protein; protein interacting with cyclin A1; Protein PROCA1
Gene Symbols PROCA1
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human PROCA1 (aa 303-352).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 147011
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.