Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PROCA1 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA554859
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554859 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554859 Supplier Thermo Scientific Supplier No. PA554859
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TTLTIERWTK EKTEPKARSW DESRCRGDCK EPDKCCWRHK QCTGHIIYPF ASDCVRHSLH LHSVNHCNCN S Highest antigen sequence identity to the following orthologs: Mouse - 35%, Rat - 37%.

PROCA1 gene ontology annotations related to this gene include arachidonic acid secretion; phospholipid metabolic process.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PROCA1
Applications Immunohistochemistry
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PROCA1
Gene Accession No. Q8NCQ7
Gene Alias 4933404M19Rik; PROCA1; proline-rich cyclin A1-interacting protein; protein interacting with cyclin A1; Protein PROCA1
Gene Symbols PROCA1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PROCA1
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 147011
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.