Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Procalcitonin Antibody (38F11), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP179065B
Description
Procalcitonin Monoclonal specifically detects Procalcitonin in Human samples. It is validated for Western Blot, ELISA, Sandwich ELISA.Specifications
Procalcitonin | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
CALCA | |
Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with fragments of human Procalcitonin conjugated with carrier protein. | |
0.2 mL | |
Cancer | |
796 | |
Human | |
Purified |
Western Blot, ELISA | |
38F11 | |
Western Blot, ELISA, Sandwich ELISA | |
ATP2B2plasma membrane calcium ATPase, ATPase, Ca++ transporting, plasma membrane 4, DKFZp686G08106, DKFZp686M088, EC 3.6.3, EC 3.6.3.8, Matrix-remodeling-associated protein 1, matrix-remodelling associated 1, MXRA1, Plasma membrane calcium ATPase isoform 4, plasma membrane calcium pump, Plasma membrane calcium pump isoform 4, plasma membrane calcium-transporting ATPase 4, PMCA4b, PMCA4PMCA4x, sarcolemmal calcium pump | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human PCT | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction