Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Procalcitonin Antibody (9D9.E11.C3.E8.F4.D4), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP24410350UG
Description
Procalcitonin Monoclonal specifically detects Procalcitonin in Human samples. It is validated for Western Blot, ELISA.Specifications
Procalcitonin | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:1000 - 1:2000, ELISA 1:100-1:2000 | |
CALC1, CALCA, Calcitonin 1, calcitonin gene-related peptide 1, CGRP, Katacalcin | |
Mouse | |
50 μg | |
796 | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot, ELISA | |
9D9.E11.C3.E8.F4.D4 | |
Unconjugated | |
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% sodium azide | |
CALCA | |
Procalcitonin Antibody was produced in mice prepared by repeated immunizations with full-length recombinant human Procalcitonin protein. | |
Primary | |
Human | |
IgG1 λ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction