Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Procalcitonin Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300233B
Description
Procalcitonin Polyclonal specifically detects Procalcitonin in Human samples. It is validated for ELISA.Specifications
Procalcitonin | |
Polyclonal | |
PBS | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
Biotin | |
CALC1, CALCA, Calcitonin 1, calcitonin gene-related peptide 1, CGRP, Katacalcin | |
Produced in rabbits immunized with purified, recombinant Procalcitonin (P01258-1; Ala26-Asn141). | |
0.1 mL | |
Cancer, Cellular Markers, Neuroscience, Phospho Specific | |
796 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction