Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    Procalcitonin Antibody, Janelia Fluor™ 549, Novus Biologicals™
 
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300233JF549
Description
Procalcitonin Polyclonal antibody specifically detects Procalcitonin in Human samples. It is validated for ELISASpecifications
| Procalcitonin | |
| Polyclonal | |
| ELISA | |
| CALC1, CALCA, Calcitonin 1, calcitonin gene-related peptide 1, CGRP, Katacalcin | |
| Produced in rabbits immunized with purified, recombinant Human Procalcitonin (Accession#: P01258-1; Ala26-Asn141) | |
| 0.1 mL | |
| Cancer, Cellular Markers, Neuroscience, Phospho Specific, Signal Transduction | |
| 796 | |
| Store at 4C in the dark. | |
| IgG | 
| ELISA | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
            -549-NBP2-24683JF549-img0001.jpg-250.jpg)