Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Procalcitonin Monoclonal Antibody (38F11)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA120292
Description
MA1-20292 detects procalcitonin in human samples.
This gene encodes the peptide hormones calcitonin, calcitonin gene-related peptide and katacalcin by tissue-specific alternative RNA splicing of the gene transcripts and cleavage of inactive precursor proteins. Calcitonin is involved in calcium regulation and acts to regulate phosphorus metabolism. Calcitonin gene-related peptide functions as a vasodilator while katacalcin is a calcium-lowering peptide. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
Procalcitonin | |
Monoclonal | |
6.1 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
P01258, P06881 | |
CALCA | |
Recombinant fragment of human procalcitonin. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Western Blot | |
38F11 | |
Unconjugated | |
CALCA | |
alpha-type CGRP; CALC1; CALCA; Calcitonin; calcitonin 1; Calcitonin carboxyl-terminal peptide; Calcitonin gene-related peptide 1; Calcitonin gene-related peptide I; calcitonin related polypeptide alpha; calcitonin/calcitonin-related polypeptide, alpha; calcitonin; calcitonin gene-related peptide 1; calcitonin-related polypeptide alpha; CCP; CGRP; CGRP1; CGRP-I; CT; Katacalcin; KC; PCT; PDN-21 | |
Mouse | |
Protein A | |
RUO | |
796 | |
4°C | |
Liquid |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction