Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Procalcitonin Monoclonal Antibody (44d9)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA120889
Description
MA1-20889 detects Procalcitonin in Human samples.
This gene encodes the peptide hormones calcitonin, calcitonin gene-related peptide and katacalcin by tissue-specific alternative RNA splicing of the gene transcripts and cleavage of inactive precursor proteins. Calcitonin is involved in calcium regulation and acts to regulate phosphorus metabolism. Calcitonin gene-related peptide functions as a vasodilator while katacalcin is a calcium-lowering peptide. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
Procalcitonin | |
Monoclonal | |
4.9 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
P01258, P06881 | |
CALCA | |
Purified, N-terminal fragment of native human Procalcitonin. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
ELISA, Western Blot | |
44d9 | |
Unconjugated | |
CALCA | |
alpha-type CGRP; CALC1; CALCA; Calcitonin; calcitonin 1; Calcitonin carboxyl-terminal peptide; Calcitonin gene-related peptide 1; Calcitonin gene-related peptide I; calcitonin related polypeptide alpha; calcitonin/calcitonin-related polypeptide, alpha; calcitonin; calcitonin gene-related peptide 1; calcitonin-related polypeptide alpha; CCP; CGRP; CGRP1; CGRP-I; CT; Katacalcin; KC; PCT; PDN-21 | |
Mouse | |
Protein A | |
RUO | |
796 | |
4°C, do not freeze | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction