Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Procalcitonin Monoclonal Antibody (4D12.D4.F3.E2.H7.E7), Rockland™
SDP

Catalog No. 501990655
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-0655 100 μg
50-199-0656 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-0655 Supplier Rockland Immunochemicals Supplier No. 200301I48
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This antibody is specific for human Procalcitonin. Cross-reactivity with Procalcitonin from other sources has not been determined.

This gene encodes the peptide hormones calcitonin, calcitonin gene-related peptide and katacalcin by tissue-specific alternative RNA splicing of the gene transcripts and cleavage of inactive precursor proteins. Calcitonin is involved in calcium regulation and acts to regulate phosphorus metabolism. Calcitonin gene-related peptide functions as a vasodilator while katacalcin is a calcium-lowering peptide. Multiple transcript variants encoding different isoforms have been found for this gene.

Specifications

Antigen Procalcitonin
Applications ELISA, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 4D12.D4.F3.E2.H7.E7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene CALCA
Gene Accession No. P01258, P06881
Gene Alias Alpha-type CGRP; CALC1; CALCA; Calcitonin; calcitonin 1; Calcitonin carboxyl-terminal peptide; Calcitonin gene-related peptide 1; Calcitonin gene-related peptide I; calcitonin related polypeptide alpha; calcitonin/calcitonin-related polypeptide, alpha; calcitonin; calcitonin gene-related peptide 1; calcitonin-related polypeptide alpha; CCP; CGRP; CGRP1; CGRP-I; CT; Katacalcin; KC; PCT; PDN-21
Gene Symbols CALCA
Host Species Mouse
Immunogen Procalcitonin Antibody was produced in mice prepared by repeated immunizations with full-length recombinant human Procalcitonin protein.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 796
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.