Learn More
Procalcitonin Monoclonal Antibody (8B5.A3.D5.D3.G6.E7), Rockland™

Mouse Monoclonal Antibody
$174.73 - $651.91
Specifications
Antigen | Procalcitonin |
---|---|
Clone | 8B5.A3.D5.D3.G6.E7 |
Concentration | 1 mg/mL |
Applications | ELISA, Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-0649
![]() ![]() |
Rockland Immunochemicals
200301I45 |
100 μg |
Each for $651.91
|
|
|||||
50-199-0650
![]() ![]() |
Rockland Immunochemicals
200301I45S |
25 μL |
Each for $174.73
|
|
|||||
Description
Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This antibody is specific for human Procalcitonin. Cross-reactivity with Procalcitonin from other sources has not been determined.
This gene encodes the peptide hormones calcitonin, calcitonin gene-related peptide and katacalcin by tissue-specific alternative RNA splicing of the gene transcripts and cleavage of inactive precursor proteins. Calcitonin is involved in calcium regulation and acts to regulate phosphorus metabolism. Calcitonin gene-related peptide functions as a vasodilator while katacalcin is a calcium-lowering peptide. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
Procalcitonin | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2 | |
Alpha-type CGRP; CALC1; CALCA; Calcitonin; calcitonin 1; Calcitonin carboxyl-terminal peptide; Calcitonin gene-related peptide 1; Calcitonin gene-related peptide I; calcitonin related polypeptide alpha; calcitonin/calcitonin-related polypeptide, alpha; calcitonin; calcitonin gene-related peptide 1; calcitonin-related polypeptide alpha; CCP; CGRP; CGRP1; CGRP-I; CT; Katacalcin; KC; PCT; PDN-21 | |
CALCA | |
IgG1 λ | |
-20°C, Avoid Freeze/Thaw Cycles | |
CALCA |
8B5.A3.D5.D3.G6.E7 | |
ELISA, Immunoprecipitation, Western Blot | |
Unconjugated | |
Mouse | |
Human | |
P01258, P06881 | |
796 | |
Procalcitonin Antibody was produced in mice prepared by repeated immunizations with full-length recombinant human Procalcitonin protein. | |
Primary | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.