Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Procalcitonin Mouse anti-Human, Clone: 14A2, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP3005231MG
Description
Procalcitonin Monoclonal specifically detects Procalcitonin in Human samples. It is validated for Western Blot, ELISA.Specifications
Procalcitonin | |
Monoclonal | |
Unconjugated | |
PBS (pH 7.4) | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, ELISA | |
14A2 | |
Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000 | |
CALC1, CALCA, Calcitonin 1, calcitonin gene-related peptide 1, CGRP, Katacalcin | |
Recombinant fragment (Human). a.a.r. 72-81 of procalcitonin (PCT) | |
1 mg | |
Signal Transduction | |
796 | |
Store at 4°C. Do not freeze. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction