Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STERIS Process NPD™ One-Step Germicidal Detergent
Shop All STERIS Corporation Products
Click to view available options
Volume (English):
1 gal.
30 gal.
55 gal.
Container Type:
Drum
Plastic Bottle
Description
- Non-sterile version of Process NPD™ family of one-step, quaternary ammonium cleaner/disinfectants made specifically for use on hard non-porous surfaces
- Made with same formulation that contains no perfumes or dyes that might interfere with critical research studies or manufacturing processes
- Use-dilution rate of 1:256
- Kill a broad spectrum of common bacteria without harming surface
How Process NPD™ One-Step Germicidal Detergent Works
- Liquid concentrate specifically formulated for disinfection of hard surfaces
- Use-dilution can be applied manually or sprayed
Why Process NPD™ One-Step Germicidal Detergent?
- Efficacy
- Quaternary ammonium active ingredients provide broad spectrum antimicrobial efficacy
- Productivity
- Formulation includes surfactants for surface cleaning while disinfecting
- Flexibility
- Ideal for use in contamination control rotation
- Reliability
- Complete lot traceability
- Assurance
- Each lot documented by certificate of analysis
Specifications
Specifications
| Product Type | Germicidal Detergent |
| pH | 10.2 to 10.7 |
| Sterility | Non-sterile |
| Composition | Quaternary ammonium |
| Container Type | Drum |
| Volume (English) | 30 gal. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction