Learn More
Description
STERIS' Process QDS Process and Research Cleaner is a phosphate-free, one-step cleaner/disinfectant formulated for use on hard non-porous surfaces. This product is recommended for use in disinfecting, sanitizing, and deodorizing hard non-porous surfaces. It is effective in hard water and organic soil loads.
HOW PROCESS QDS PROCESS AND RESEARCH CLEANER WORKS
STERIS' Process QDS Process and Research Cleaner is a liquid concentrate specifically formulated for the disinfection of hard non-porous surfaces. The use-dilution can be applied manually or sprayed.
WHY PROCESS QDS PROCESS AND RESEARCH CLEANER?
- Efficacy - Quaternary ammonium active ingredients provide broad spectrum antimicrobial efficacy.
- Productivity - Formulation includes surfactants for surface cleaning while disinfecting.
- Reliability - Ideal for use in a contamination control rotation.
- Flexibility - Can be used as a disinfectant or sanitizer.
- Assurance - Complete lot traceability with each lot documented by Certificate of Analysis.
Specifications
Specifications
| Product Type | Cleaner/Disinfectant |
| Form | Clear Liquid |
| pH | 6.1-8.8 (Undiluted) |
| Scent | Unscented |
| Sterility | Non-sterile |
| Composition | Quaternary ammonium |
| Container Type | Drum |
| Size | 55 gal. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.