Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chembio Procise IFX Assay Infliximab
SDP

Catalog No. 21010243
Change view
Click to view available options
Form:
Freeze-dried
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Form
21-010-243 Freeze-dried
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 21-010-243 Supplier Chembio Supplier No. 5385
Add to Cart
Add to Cart
This item is not returnable. View return policy

Procise IFX™ Assay enables rapid, quantitative monitoring of infliximab in serum using the ProciseDx™ Analyzer, it supports IBD treatment decisions with results in under 5 minutes.

  • Quantitative TDM for Infliximab: Enables rapid measurement of infliximab levels in serum to support therapeutic management of IBD patients
  • Fast Results: Delivers accurate, quantitative results in under 5 minutes using the ProciseDx™ Analyzer
  • Validated for Clinical Use: FDA-cleared for use in moderate-complexity labs; supports both adult and pediatric Crohn's disease and ulcerative colitis care
  • Proven Analytical Performance: Sensitive and specific TR-FRET-based immunoassay with a reportable range of 1.2 to 50 μg/mL
  • Simple Workflow: Uses standard serum samples and minimal hands-on time

Specifications

Certifications/Compliance FDA DeNovo
Content And Storage 15°C to 30°C (59°F to 86°F)
Control Sets Included High and Low
CE Marker Yes
Clia Complexity Moderate (Prescription Use Only)
Detectable Analytes Infliximab
Detection Method Measures Relative Fluorescence Units (RFU)
DoA Calibrators No
Sample Type Serum
Type Test Cartridge
Disposable Single-use
For Use With (Equipment) ProciseDx Analyzer
Includes 20 Reagent Cartridges and Buffer Bulbs, 4 Controls a 2- High and 2- Low, Product Insert
Level High, Low
Quantity 20 Pk.
Range 1.2 to 50.0 μg/mL
Sample Size 20 μL
Analysis Time <5 minutes
Dimensions (L x W x H) 15 x 8 x 5.7 in.
Form Freeze-dried
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.