Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

proCNP Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501980965
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0965 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0965 Supplier Bioss Supplier No. BS1069R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NPPC (Natriuretic Peptide C) is a hormone which plays a role in endochondral ossification through regulation of cartilaginous growth plate chondrocytes proliferation and differentiation. The protein may also be vasoactive and natriuretic. It specifically binds and stimulates the cGMP production of the NPR2 receptor, and also binds the clearance receptor NPR3.

Specifications

Antigen proCNP
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene NPPC
Gene Accession No. P23582
Gene Alias BOS_2151; Cnp; CNP2; CNP-22; CNP-29; CNP-53; C-type natriuretic peptide; C-type natriuretic peptide/factor; INT3; lbab; LOC100344147; natriuretic peptide C; natriuretic peptide precursor C; natriuretic peptide precursor type C; natriuretic peptide type C; NOTCH3; NPPC; SVSP15
Gene Symbols NPPC
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human proCNP.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4880
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.