Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ ProCoat™ Cold Resistant PVC Gloves with Detachable Terry Liner

Catalog No. 19046390 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
2X-Large
Large
X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-046-390 X-Large
19-046-389 Large
19-046-391 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-046-390 Supplier PIP™ Supplier No. 588658DL/XL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Foam insulation combined with a cold-resistant outer shell helps to keep hands warm and dry. A detachable, 7-gauge insulating liner provides the option for additional thermal protection.

  • Cold-resistant outer shell
  • Foam insulation helps keep hands warm and dry
  • Seamless knit construction protects hands without sacrificing comfort or dexterity
  • Sandy finish enhances grip in wet and slippery conditions
  • Detachable, insulating 7-gauge terry cloth liner
  • Ideal for handling grease, oils, acids, caustics and solvents in cold environments
  • Liquid-proof

Specifications

Coating Material PVC
Color Blue
Exterior Finish Fully Textured
Hand Specific
Material PVC
Product Type Cold-resistant Gloves
Size X-Large
For Use With (Application) Cold Environments, Construction, Solvent Handling, and Warehousing
Glove Cuff Style Open
Liner Material Cotton
Series 58-8658DL
Yarn Gauge 7
Length (English) 12 in.
Length (Metric) 30.5 cm
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.