Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Procollagen I C-Peptide Monoclonal Antibody (8), Invitrogen™

Catalog No. BTE0020802
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BTE0020802 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BTE0020802 Supplier Thermo Scientific Supplier No. BTE0020802

Mouse Monoclonal Antibody

BTE 002-08-02 is specific for native PICP from human amniotic fluid. Crossreactivity to other species is not determined. BTE 002-08-02 has been successfully used in ELISA procedures. BTE 002-08-02 is especially well suited as the catching antibody in a sandwich ELISA in combination with BTE 002-18-02. The BTE 002-08-02 epitope differs from that of BTE 002-18-02. NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL

Collagen type I is present in soft connective tissue and bone where it represents approximately 90% of the organix matrix. Collagen type I is produced from procollagen when two teloterminal propeptides are digested from the procursor and released into the circulation. The propeptide levels can be measured in serum as indicators of collagen production. Elevated values of these propeptides in serum are found in patients with various bone metabolism defects. Procollagen type I C-terminal propeptide (PICP) has a molecular weight of approximately 100 kDa and the normal range in serum depends on age and sex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Procollagen I C-Peptide
Applications ELISA
Classification Monoclonal
Clone 8
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 15mM sodium azide; pH 7.4
Gene COL1A1
Gene Accession No. P02452
Gene Alias Alpha-1 type I collagen; alpha1(I) procollagen; COL1A1; collagen alpha 1 chain type I; collagen alpha-1(I) chain; collagen alpha-1(I) chain preproprotein; collagen of skin, tendon and bone, alpha-1 chain; collagen type I alpha 1; collagen, type I, alpha 1; C-terminal Collagen; EDSC; OI1; OI2; OI3; OI4; PICP; pro-alpha-1 collagen type 1; Procol; Procollagen; type I proalpha 1; type I procollagen alpha 1 chain
Gene Symbols COL1A1
Host Species Mouse
Immunogen PICP purified from second trimester amniotic fluid by affinity chromatography.
Purification Method Protein A/G
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1277
Target Species Human
Content And Storage 4° C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.