Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Procollagen I C-Peptide Polyclonal Antibody, Invitrogen™
GREENER_CHOICE

Catalog No. FEPA5145235
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5145235 200 μL
FEPA5145236 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FEPA5145235 Supplier Invitrogen™ Supplier No. PA5145235
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Collagen type I is present in soft connective tissue and bone where it represents approximately 90% of the organix matrix. Collagen type I is produced from procollagen when two teloterminal propeptides are digested from the procursor and released into the circulation. The propeptide levels can be measured in serum as indicators of collagen production. Elevated values of these propeptides in serum are found in patients with various bone metabolism defects. Procollagen type I C-terminal propeptide (PICP) has a molecular weight of approximately 100 kDa and the normal range in serum depends on age and sex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Procollagen I C-Peptide
Applications Immunohistochemistry (Paraffin), Western Blot, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
Gene COL1A1
Gene Accession No. P02452
Gene Alias Alpha-1 type I collagen; alpha1(I) procollagen; COL1A1; collagen alpha 1 chain type I; collagen alpha-1(I) chain; collagen alpha-1(I) chain preproprotein; collagen of skin, tendon and bone, alpha-1 chain; collagen type I alpha 1; collagen, type I, alpha 1; C-terminal Collagen; EDSC; OI1; OI2; OI3; OI4; PICP; pro-alpha-1 collagen type 1; Procol; Procollagen; type I proalpha 1; type I procollagen alpha 1 chain
Gene Symbols COL1A1
Host Species Rabbit
Immunogen KLH conjugated Human CTXI protein, E1207-A1218 (Accession #Q9NPZ5).
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1277
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.