Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Procollagen I N-Peptide Monoclonal Antibody (C6)
GREENER_CHOICE

Catalog No. PIMA541662
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA541662 200 μL
PIMA543883 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA541662 Supplier Invitrogen™ Supplier No. MA541662
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Procollagen I N-Peptide Monoclonal Antibody for Western Blot, IHC (P)

Procollagen type I N-terminal propeptide (P1NP) is a specific marker of bone formation, reflecting the synthesis of type I collagen, which constitutes the major part of the bone matrix. P1NP is derived during the conversion of procollagen to collagen, produced by osteoblasts during bone formation. Its primary clinical application lies in the diagnosis and monitoring of metabolic bone diseases, particularly osteoporosis. In osteoporotic patients, P1NP levels can predict bone mineral density (BMD) responses to treatments like teriparatide and romosozumab. Low serum levels of P1NP are indicative of a high risk of BMD loss, providing a valuable tool for premenopausal women with systemic lupus erythematosus (SLE) and other conditions affecting bone health.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Procollagen I N-Peptide
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone C6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene COL1A1
Gene Accession No. P02452, P02454
Gene Alias P1NP; Procollagen I N
Gene Symbols COL1A1
Host Species Mouse
Immunogen Recombinant protein Procollagen I N-Terminal Propeptide.
Purification Method Protein A/G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1277, 29393
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.