Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Procollagen I N-Peptide Monoclonal Antibody (C8), Invitrogen™

Catalog No. FEMA551184
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA551184 20 μL
FEMA551183 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FEMA551184 Supplier Invitrogen™ Supplier No. MA551184
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Procollagen type I N-terminal propeptide (P1NP) is a specific marker of bone formation, reflecting the synthesis of type I collagen, which constitutes the major part of the bone matrix. P1NP is derived during the conversion of procollagen to collagen, produced by osteoblasts during bone formation. Its primary clinical application lies in the diagnosis and monitoring of metabolic bone diseases, particularly osteoporosis. In osteoporotic patients, P1NP levels can predict bone mineral density (BMD) responses to treatments like teriparatide and romosozumab. Low serum levels of P1NP are indicative of a high risk of BMD loss, providing a valuable tool for premenopausal women with systemic lupus erythematosus (SLE) and other conditions affecting bone health.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Procollagen I N-Peptide
Applications Immunohistochemistry (Paraffin), Western Blot, Western Blot
Classification Monoclonal
Clone C8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
Gene COL1A1
Gene Accession No. P02452, P02454
Gene Alias P1NP; Procollagen I N
Gene Symbols COL1A1
Host Species Mouse
Immunogen KLH conjugated Human PINP protein (Accession #P02452).
Purification Method Protein A/G
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1277, 29393
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.