Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Procollagen IIA N-Peptide Monoclonal Antibody (02)
GREENER_CHOICE

Catalog No. BTE0030202
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BTE0030202 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BTE0030202 Supplier Invitrogen™ Supplier No. BTE0030202
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BTE 003-02-02 is specific human PIIANP. BTE 003-02-02 has been successfully used ELISA, and Immunohistochemistry procedures. Epitope is located at the amino-terminal part of procollagen type IIA. NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL.

Collagen type II is the most abundant protein of cartilage matrix. Two isoforms exist and collagen type IIA is involved in early development of cartilage in embryos. The collagen is produced from type II procollagen when two teloterminal propeptides are digested from the procursor and released into the circulation. Procollagen type II N-terminal propeptide (PIIANP) is considered a marker of cartilage synthesis and the normal concentration in serum is approximately 30 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Procollagen IIA N-Peptide
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 02
Concentration 1.02 mg/mL
Conjugate Unconjugated
Formulation PBS with 15mM sodium azide; pH 7.4
Gene COL2A1
Gene Accession No. P02458
Gene Alias alpha-1 type II collagen; ANFH; AOM; arthroophthalmopathy, progressive (Stickler syndrome); cartilage collagen; chondrocalcin; COL11A3; COL2A1; Collagen alpha-1(II) chain; collagen II, alpha-1 polypeptide; collagen type II alpha 1 chain; collagen, type II, alpha 1; SEDC; STL1
Gene Symbols COL2A1
Host Species Mouse
Immunogen Recombinant expressed PIIANP.
Purification Method Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1280
Target Species Human
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.