Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Procollagen Type I C-Peptide (PIP) Monoclonal Antibody (PC8-7), QED Bioscience™

Mouse Monoclonal Antibody
Supplier: Leinco Technologies Inc 42043100UG

This item is not returnable.
View return policy
Description
Procollagen Type I C-Peptide (PIP) Monoclonal antibody specifically detects Procollagen Type I C-Peptide (PIP) in Bovine, Canine, Equine, Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Western BlotSpecifications
Procollagen Type I C-Peptide (PIP) | |
Monoclonal | |
0.5-1.0 mg/mL | |
PBS with 1% BSA and no preservative; pH 7.4 | |
P02452, P02453, Q9XSJ7 | |
Mouse | |
Protein A | |
RUO | |
100033877, 1277, 282187, 403651 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
PC8-7 | |
Unconjugated | |
COL1A1 | |
Alpha-1 type I collagen; alpha1(I) procollagen; COL1A1; collagen alpha 1 chain type I; collagen alpha-1(I) chain; collagen alpha-1(I) chain preproprotein; collagen of skin, tendon and bone, alpha-1 chain; collagen type I alpha 1; collagen, type I, alpha 1; C-terminal Collagen; EDSC; OI1; OI2; OI3; OI4; PICP; pro-alpha-1 collagen type 1; Procol; Procollagen; type I proalpha 1; type I procollagen alpha 1 chain | |
Human Procollagen Type I C-Peptide | |
100 μg | |
Primary | |
Bovine, Canine, Equine, Human | |
Antibody | |
IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction