Learn More
PROCR Mouse anti-Human, Clone: OTI3E7, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF805524

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene is a receptor for activated protein C, a serine protease activated by and involved in the blood coagulation pathway. The encoded protein is an N-glycosylated type I membrane protein that enhances the activation of protein C. Mutations in this gene have been associated with venous thromboembolism and myocardial infarction, as well as with late fetal loss during pregcy.Specifications
PROCR | |
Monoclonal | |
Unconjugated | |
PROCR | |
CCCA, CCD41, EPCR | |
Mouse | |
Affinity Chromatography | |
RUO | |
10544 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI3E7 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9UNN8 | |
PROCR | |
Human recombit protein fragment corresponding to amino acids 1-210 of human PROCR produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.