Learn More
PRODH Mouse anti-Human, Clone: OTI1H6, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF810559

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene is a mitochondrial proline dehydrogenase that catalyzes the first step in proline degradation. Defects in this gene are a cause of hyperprolinemia type 1 and possibly susceptibility to schizophrenia 4 (SCZD4). The gene is located on chromosome 22q11.21, a region which has also been associated with the contiguous gene deletion syndromes DiGeorge syndrome and CATCH22 syndrome.Specifications
PRODH | |
Monoclonal | |
Unconjugated | |
PRODH | |
HSPOX2, PIG6, POX, PRODH1, PRODH2, TP53I6 | |
Mouse | |
Affinity Chromatography | |
RUO | |
5625 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI1H6 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
O43272 | |
PRODH | |
Full length human recombit protein of human PRODH produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.