Learn More
Invitrogen™ PRODH Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5120955
Description
Positive test controls include: Mouse liver, Mouse kidney, Mouse heart, Rat liver, Rat kidney. The target is usually found in the following locations: Mitochondrion matrix. Immunogen sequence: IDSRTKLSKH LVVPNAQTGQ LEPLLSRFTE EEELQMTRML QRMDVLAKKA TEMGVRLMVD AEQTYFQPAI SRLTLEMQRK FNVEKPLIFN TYQCYLKDAY DNVTLDVELA RREGWCFGAK LVRGAYLAQE RARAAEIGYE DPINPTYEAT NAMYHRCLDY VLEELKHNAK AKVMVASHNE DTVRFALRRM EELGLHPADH QVYFGQLLGM CDQISFPLGQ AGYPVYKYVP YGPVMEVLPY LSRRALENSS LMKGTHRERQ LLWLELLRRL RTGNLFHRPA.
The protein encoded by this gene is a mitochondrial proline dehydrogenase that catalyzes the first step in proline degradation. Defects in this gene are a cause of hyperprolinemia type 1 and possibly susceptibility to schizophrenia 4 (SCZD4). The gene is located on chromosome 22q11.21, a region which has also been associated with the contiguous gene deletion syndromes DiGeorge syndrome and CATCH22 syndrome.
Specifications
PRODH | |
Polyclonal | |
Unconjugated | |
Prodh | |
HSPOX2; p53-induced gene 6 protein; PIG6; POX; POX2; Pro1; Pro-1; Prodh; PRODH 1; PRODH 2; PRODH1; PRODH2; proline dehydrogenase; proline dehydrogenase (oxidase) 1; proline dehydrogenase (proline oxidase); proline dehydrogenase 1; proline dehydrogenase 1, mitochondrial; proline dehydrogenase, mitochondrial; proline oxidase; Proline oxidase 2; proline oxidase, mitochondrial; SCZD4; TP53I6; tumor protein p53 inducible protein 6; Ym24d07 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
19125, 5625, 680409 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.86 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O43272, Q9WU79 | |
Prodh, Prodh1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 321-600 of human PRODH (NP_057419.4). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.