Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRODH Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986826
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6826 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6826 Supplier Bioss Supplier No. BS5813R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene is a mitochondrial proline dehydrogenase that catalyzes the first step in proline degradation. Defects in this gene are a cause of hyperprolinemia type 1 and possibly susceptibility to schizophrenia 4 (SCZD4). The gene is located on chromosome 22q11.21, a region which has also been associated with the contiguous gene deletion syndromes DiGeorge syndrome and CATCH22 syndrome.

Specifications

Antigen PRODH
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Prodh
Gene Accession No. O43272
Gene Alias HSPOX2; p53-induced gene 6 protein; PIG6; POX; POX2; Pro1; Pro-1; Prodh; PRODH 1; PRODH 2; PRODH1; PRODH2; proline dehydrogenase; proline dehydrogenase (oxidase) 1; proline dehydrogenase (proline oxidase); proline dehydrogenase 1; Proline dehydrogenase 1, mitochondrial; proline dehydrogenase, mitochondrial; Proline oxidase; Proline oxidase 2; proline oxidase, mitochondrial; SCZD4; TP53I6; tumor protein p53 inducible protein 6; Ym24d07
Gene Symbols Prodh
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human PRODH.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5625
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.