Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PRODH2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542039
Description
Peptide sequence: LGIPLDGTVC FGQLLGMCDH VSLALGQAGY VVYKSIPYGS LEEVIPYLIR Sequence homology: Cow: 100%; Dog: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rat: 100%.
PRODH2 is similar to proline dehydrogenase (oxidase) 1, a mitochondrial enzyme which catalyzes the first step in proline catabolism. The function of this protein has not been determined.The protein encoded by this gene is similar to proline dehydrogenase (oxidase) 1, a mitochondrial enzyme which catalyzes the first step in proline catabolism. The function of this protein has not been determined.
Specifications
PRODH2 | |
Polyclonal | |
Unconjugated | |
PRODH2 | |
2510028N04Rik; 2510038B11Rik; HSPOX1; Hydroxyproline dehydrogenase; HYPDH; Kidney and liver proline oxidase 1; MmPOX; MmPOX1; PO; POX1; Probable proline dehydrogenase 2; probable proline oxidase 2; PRODH2; proline dehydrogenase (oxidase) 2; proline dehydrogenase 2; proline oxidase 1; proline oxidase-like protein | |
Rabbit | |
Affinity chromatography | |
RUO | |
58510 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9UF12 | |
PRODH2 | |
Synthetic peptide directed towards the C-terminal of human PRODH2 (aa 451-500). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction