Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ ProDots Human IGF-I Recombinant Protein

Catalog No. p-7083352
Click to view available options
Quantity:
250 μg
50 μg
This item is not returnable. View return policy
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The ProDots Recombinant Human IGF-I/IGF-1 Protein is derived from E. coli. The ProDots Recombinant Human IGF-I/IGF-1 Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number P05019
For Use With (Application) Bioactivity
Gene ID (Entrez) 3479
Molecular Weight (g/mol) Observed M.W.: 7kDa, reducing conditions, Theoretical M.W.: 7.6kDa
Name IGF-I/IGF-1
Quantity 50 μg
Source E. coli-derived human IGF-I/IGF-1 protein Gly49-Ala118
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt at room temperature as supplied. 12 months from date of receipt at 2-8° C as supplied. 1 month at 2-8° C under sterile conditions after reconstitution. 3 months at -20 to -80° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias IBP1, IGF1, IGF-1, IGF1A, IGFI, IGF-I, IGF-IA, IGF-IB, insulin-like growth factor 1, insulin-like growth factor 1 (somatomedin C), insulin-like growth factor I, insulin-like growth factor IA, insulin-like growth factor IB, Mechano growth factor, MGF, Somatomedin A, Somatomedin C, Somatomedin-C
Common Name IGF-I
Gene Symbol IGF1
Biological Activity Measured in a serum-free cell proliferation assay using MCF-7 human breast cancer cells. Karey, K.P. et al. (1988) Cancer Research 48:4083. The ED50 for this effect is typically 0.3-1.5ng/mL.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Reconstitution For a stock solution, reconstitute at 200 μg/mL in sterile PBS, or simply roll ProDot™ directly into cell culture medium for immediate use.
Expression System E. coli-derived Gly49 - Ala118
Purity or Quality Grade >97%, by SDS-PAGE under reducing conditions and visualized by silver stain
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.