Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ ProDots Recombinant Human Sonic Hedgehog/Shh (C24II) Protein
Click to view available options
Quantity:
250 μg
5 μg
50 μg
Description
The ProDots Recombinant Human Sonic Hedgehog/Shh (C24II) Protein is derived from E. coli. The ProDots Recombinant Human Sonic Hedgehog/Shh (C24II) Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Accession Number | NP_000184 |
| Characterization | Bioactivity |
| For Use With (Application) | Bioactivity |
| Gene ID (Entrez) | 6469 |
| Molecular Weight (g/mol) | 20kDa |
| Quantity | 250 μg |
| Source | E. coli-derived human Sonic Hedgehog/Shh protein Cys24-Gly197 (Cys24lle-lle),with N-terminal Met |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt at room temperature as supplied. 12 months from date of receipt at 2-8° C as supplied. 1 month at 2-8° C under sterile conditions after reconstitution. 3 months at -20 to -80° C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.01EU per 1μg of the protein by the LAL method. |
| Gene Alias | HHG1, HHG-1, HLP3, HPE3, MCOPCB5, MCOPCB5sonic hedgehog (Drosophila) homolog, Shh, ShhNC, SMMCI, SMMCIsonic hedgehog homolog (Drosophila), sonic hedgehog, sonic hedgehog homolog, sonic hedgehog protein, TPT, TPTPS |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction