Learn More
Thermo Scientific™ Productainer™ BioProcess Container (BPC), 100L with 4 Top Ports, 3D Single-Use BPC

Description
Using Thermo Scientific™ Productainer™ BPC bag with 4 top ports, package, store, and transport 50 to 200L volumes of media, buffers, water for injection (WFI), process liquids, and biological products.
Highlights:
- Three sizes - 50, 100 and 200L
- Film optimized for flexibility and durability: superior stress resistance, puncture resistance and physical barrier properties to ensure stability of contents
- Gamma-irradiation sterilized
- Squared tube design with polyethylene ports located on the top of the BPC for easy access
- Space-efficient, ergonomic alternative to rigid walled carboys and steel tanks
- Customizable via the industry's largest library of qualified components
Recommended for:
Harvest - Collection and storage of bulk product from bioreactors
Storage - Sterile storage of media, buffers and process intermediates
Separation - Feeding and receiving of sterile liquids from separation systems
Purification - Buffer feeding to purification system, fraction collection and storage
Filling - Sterile reservoir for filling systems
Transport and Shipping - Bulk sterile liquids, buffers, media and process intermediates within and between facilities
Specifications
Specifications
| Green Features | Fewer resources used and less waste |
| Capacity (Metric) | 100 L |
| Description | 100 L dip tube: 71 cm (28 in.) |
| Film Description | CX5-14 |
| Port Detail | Top Ports: 2 x 0.25 in. 2 x 0.38 in. |
| For Use With (Equipment) | SV50076.03 |
| Material | Polyethylene |
| Outer Packaging | Packages placed in strong cardboard cartons with product and lot identification |
| Protective Inner Packaging | Packed and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area |
| Quantity | 100 L |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.